Learn More
LGC Genomics Llc (Lucigen) Endura Chemically Competent Cells, 12 Rxns (SOLOs)Clone and maintain unstable or repetitive sequences and lentiviral libraries with high efficiency. Ideal for GeCKO CRISPR library transformation.
Catalog No. NC1419830
Encompass
Lucigens Endura Competent Cells are a commonly used strain for cloning sequences that suffer unwanted recombination events in other strains. Clones with inverted repeats or other sequences prone to recombination are commonly found in retroviral genes, and require cells such as Endura to be propagated stably.
Catalog No. NC1419830 Supplier LGC Genomics Llc Supplier No. 602411
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Lucigens Endura Competent Cells are a commonly used strain for cloning sequences that suffer unwanted recombination events in other strains. Clones with inverted repeats or other sequences prone to recombination are commonly found in retroviral genes, and require cells such as Endura to be propagated stably.