Learn More
Lonza Walkersville Inc Endotoxin Challenge Vials
Supplier:  Lonza Walkersville Inc 00192568
Encompass
UPDATE (May 2020):We are discontinuing the performance of a sterility test for our Endotoxin Challenge Vials. As part of the production process, the stoppers are autoclaved prior to use.Additionally, all glassware is depyrogenated at extremely high temperatures well in excess of standard depyrogenation process conditions. These conditions provide for ample assurance that the vials and stoppers are suitable for production of Endotoxin Challenge Vials. Post filling, the product is lyophilized. Since there is no remaining water, conditions would not supportgrowth of any organism to degrade the product.Therefore, there is no added value or additional quality assurance in performing a sterility test after production.For use in oven validation studies, contains > 1,000 EU/vial. The vials may be tested using any of our endotoxin detection kits.Benefits:Does not contain fillerApplications:For oven validation studiesContent:> 1000 EU/Vial
Catalog No. NC0009125
Only null left