missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Endogenous Biotin-Blocking Kit

Catalog No. E21390
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E21390 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E21390 Supplier Invitrogen™ Supplier No. E21390

missing translation for 'validMainframeMsg'

Endogenous Biotin-Blocking Kit

The Endogenous Biotin-Blocking Kit provides a method and reagents for a pretreatment that reduces or eliminates background signals when biotin-avidin or biotin-streptavidin detection systems are used to identify cellular targets.

The Endogenous Biotin-Blocking Kit offers:
  • Improved results—increased signal-to-noise ratio facilitates cell target identification
  • Ease of use—pipette-free dropper bottle offers speed and convenience
  • Fast—endogenous background reduction in under an hour
  • Flexibility—the kit can be used to help improve results from conventional immunohistochemistry, immunocytochemistry, RNA FISH, and ELISA procedures

    Biotin occurs naturally, functioning as an enzyme cofactor in the cytosol and mitochondria of a wide variety of cell types. The Endogenous Biotin-Blocking Kit minimizes interference from the endogenous biotin found in enzymes of mammalian cells and tissues, through a two-step blocking strategy. First, an excess of unlabeled streptavidin is added to the specimen to bind endogenous biotin-rich enzymes. The streptavidin is subsequently blocked with an excess of unlabeled biotin, effectively rendering the cell or tissue sample free of available biotin-binding sites.
  • Specifications

    Type Endogenous Biotin-Blocking Kit
    Reagent Type Blocking⁄Background Suppression Reagent
    Content And Storage Store in refrigerator at 2°C to 8°C
    Quantity 1 kit
    Shipping Condition Room Temperature

    For Research Use Only. Not for use in diagnostic procedures.