missing translation for 'onlineSavingsMsg'
Learn More

endo-N-Hydroxy-5-norbornene-2,3-dicarboximide, 97%

Catalog No. p-7021038
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Catalog No. Quantity
AAA1320509 10 g
AAA1320518 50 g
AAA1320530 250 g
3 options

Catalog No. AAA1320509

Supplier: Thermo Scientific Chemicals A1320509

Only null left

CAS: 21715-90-2 | C9H9NO3 | 179.18 g/mol

It decreases racemization in peptide synthesis and inhibits formation of N-acylureas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It decreases racemization in peptide synthesis and inhibits formation of N-acylureas.

Solubility
Soluble in ethanol

Notes
Store at room temperature. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 21715-90-2
Molecular Formula C9H9NO3
Molecular Weight (g/mol) 179.18
MDL Number MFCD00065691
InChI Key ZUSSTQCWRDLYJA-UHFFFAOYNA-N
Synonym honb, n-hydroxy-5-norbornene-2,3-dicarboximide, 2-hydroxy-3a,4,7,7a-tetrahydro-1h-4,7-methanoisoindole-1,3 2h-dione, 4-hydroxy-4-azatricyclo 5.2.1.0 2,6 dec-8-ene-3,5-dione, n-hydroxynorborn-5-ene-2,3-dicarboximide, n-hydroxy-5-norbornene-2,3-dicarboxylic imide, 5-norbornene-2, n-hydroxy, 4,7-methano-1h-isoindole-1,3 2h-dione, 3a,4,7,7a-tetrahydro-2-hydroxy, 4,3 2h-dione, 3a,4,7,7a-tetrahydro-2-hydroxy, 4-hydroxy-4-azatricyclo 5.2.1.0<2,6> dec-8-ene-3,5-dione
PubChem CID 89529
IUPAC Name 4-hydroxy-4-azatricyclo[5.2.1.0²,⁶]dec-8-ene-3,5-dione
SMILES ON1C(=O)C2C3CC(C=C3)C2C1=O

Specifications

Melting Point 166°C to 170°C
Quantity 10 g
Beilstein 13540
Solubility Information Soluble in ethanol
Formula Weight 179.18
Percent Purity 97%
Chemical Name or Material endo-N-Hydroxy-5-norbornene-2,3-dicarboximide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 244-538-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only