Endoglycosidase H is a recombinant glycosidase which cleaves within the chitobiose core of high mannose and some hybrid oligosaccharides from N-linked glycoproteins.
Catalog No.
50811826
Supplier
New England Biolab LtdSupplier No.
P0702S
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Endoglycosidase H is a recombinant glycosidase which cleaves within the chitobiose core of high mannose and some hybrid oligosaccharides from N-linked glycoproteins.