missing translation for 'onlineSavingsMsg'
Learn More

Encon™ Shower Head Assembly

Catalog No. 296354901
Change view
Click to view available options
Material:
ABS Plastic
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
296354901 ABS Plastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 296354901 Supplier Encon™ Supplier No. 01 0540 03
Only null left

30 GPM yellow ABS shower head

  • 20.32cm (8 in.)
  • Galvanized, stainless-steel pipe
  • Polane™-coated, corrosion-resistant
  • PVC corrosion-resistant
  • 2.54cm (1 in.) connection

Specifications

Color Yellow
For Use With (Equipment) Shower and Combination Eye and Face Wash
Material ABS Plastic
Product Type Shower Head