missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Enalapril maleate salt

Catalog No. AAJ6393206
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6393206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6393206 Supplier Thermo Scientific Chemicals Supplier No. J6393206

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 76095-16-4492.53 g/mol

Enalapril maleate salt is used as a long-acting ACE inhibitor and antihypertensive, used to study diabetic angiopathy in diabetic rats and inhibition of ACE in hog plasma (I50=1.2nM). ACE inhibitors disturb the enin-angiotensin- aldosterone system. Additional ACE inhibitors include: Enalapril, Enalaprilat dihydrate, Enalaprilat, and Enalapril-d5 Maleate Salt, among others.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Enalapril maleate salt is used as a long-acting ACE inhibitor and antihypertensive, used to study diabetic angiopathy in diabetic rats and inhibition of ACE in hog plasma (I50=1.2nM). ACE inhibitors disturb the enin-angiotensin- ald

Chemical Identifiers

CAS 76095-16-4

Specifications

Color White
Content And Storage Store at -20°C
Format Powder
Molecular Weight (g/mol) 492.53
Product Type Enalapril maleate salt
Quantity 5 g

RUO – Research Use Only