Learn More
Thermo Scientific™ Empty Vial
Catalog No. R21922
Change view
Click to view available options
Quantity:
120 Vials/Pk.
Catalog No. Quantity
R21922 120 Vials/Pk.
1 options
Catalog No. R21922 Supplier Thermo Scientific™ Supplier No. RE21922
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Collect and transport fecal samples for enteric bacterial test procedures not compatible with fecal preservatives, complete with industry standard color-coded, sporked caps.
Collect and transport fecal samples for enteric bacterial test procedures not compatible with fecal preservatives. Thermo Scientfic™ Empty Vial is part of a complete collection of quality fecal transports which come with industry standard color-coding and easy-to-use sporked caps.
Add convenience and safety to your fecal collection procedures with a broad line of transport products which have a long shelf life and are designed especially for enteric diagnostic testing. Look to us for convenient kit packaging options, including bulk kits. Multilingual patient collection instructions are included. Choose from a wide range of collection and preservation products for fecal specimens, which is important to the successful growth, isolation and detection of microorganisms.
  • Cary Blair with Indicator
  • Formalin 10%
  • Formalin 5%
  • PVA Modified Fixative
  • SAF Fixative
  • Zinc PVA Fixative
  • Empty Vial
Not all products are available for sale in all territories. Please inquire. Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Product Type Fecal Transport Empty Vial
Description Empty Vial
Fill Volume 15 mL
Packaging Single vial. Packaged in partitioned boxes.
Quantity 120 Vials/Pk.