missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Empty SPE Tube (No Frits)

Catalog No. 111016136
Change view
Click to view available options
Volume (Metric):
1 mL
3 mL
12 mL
20 mL
60 mL
Quantity:
108/Pk.
12/Pk.
20/Pk.
54/Pk.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Quantity
111016136 1 mL 108/Pk.
111010875 3 mL 54/Pk.
111011429 12 mL 20/Pk.
111011428 20 mL 12/Pk.
111011374 60 mL 12/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 111016136 Supplier MilliporeSigma™ Supelco™ Supplier No. 57240U

Empty SPE Tubes allow you to pack your own SPE cartridges

When used in conjunction with SPE Tube Adapters, empty SPE tubes can be stacked on top of a standard SPE cartridge during processing. The empty SPE cartridge acts as a reseroir to accommodate larger sampling volumes.

Specifications

Column Type SPE
Description Empty
For Use With (Application) Food and Beverages
Type Solid Phase Extraction Column
Volume (Metric) 1 mL
Material Polypropylene
Quantity 108/Pk.