missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Empty FastPrep™ 2 mL Lysing Matrix Tubes

Catalog No. mp115076600
Change view
Click to view available options
Volume (Metric):
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
MP115076600 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP115076600 Supplier MP Biomedicals™ Supplier No. 115076600
Only null left

Available empty for those who wish to fill their own tubes. Empty FastPrep™ 2 mL Lysing Matrix Tubes (non-skirted, no caps) are 2 mL tubes that work with FastPrep or other bead-beater instruments to lsye, grind or homogenize samples.

  • Used in FastPrep systems.
  • Add sample to empty tube, add appropriate Lysing Matrix material and lysing buffer, and apply cap.
  • Empty FastPrep 2 mL tubes are screw cap tubes and can be used with any color cap.
  • They are designed for grinding and lysing samples with FastPrep instruments.
  • Non-skirted

Specifications

Color Clear
Volume (Metric) 2 mL
Storage Requirements Room Temperature (15°C to 30°C)
Quantity 5000 x 2 mL
Product Type Empty Lysing Matrix Tubes

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.