missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Empty FastPrep Tubes (Caps Not Included)
Description
- EMPTY FASTPREP Tube is used in Fastprep system, to hold sample for Homogenization.
- Empty FastPrep tubes are 2 mL volume and are the tubes that are used to manufacture all MP Bio Lysing Matrix Tubes.
- They are available for purchase empty for those who wish to fill their own tubes.
- Use with color-coded Caps.
- Add sample to empty tube, add appropriate Lysing Matrix material and lysing buffer, and apply cap. Process using FastPrep instrument or other bead-beater to lsye, grind or homogenize sample.
Specifications
Specifications
| Color | Natural |
| Description | EMPTY FASTPREP™ Tube is used in Fastprep system, to hold sample for Homogenization. |
| Volume (Metric) | 2 mL |
| Storage Requirements | Room Temperature (15 to 30°C) |
| Quantity | 1000 x 2 mL |
| Format | Tube |