Learn More
Thermo Scientific™ Empty Centrifuge Tubes for QuEChERS
Empty Thermo Scientific™ Centrifuge Tubes for QuEChERS sample preparation are ideal for handling your QuEChERS samples. They come with screw caps for secure sealing, and are packaged separately.
Supplier: Thermo Scientific™ 60106CT50
Description
Thermo Scientific™ Empty Centrifuge Tubes for QuEChERS sample preparation provide a reliable solution for your sample preparation needs. Featuring screw caps, these tubes help ensure secure sealing, and are designed for easy handling to fit your centrifugation needs.
- Compatible with QuEChERS sample preparation
- For use in centrifuges
- Secure sealing with screw caps
- Ideal for precise and efficient analysis of pesticides in foods
Specifications
| Polypropylene | |
| QuEChERS | |
| 50 mL | |
| Blue screw cap |
| Empty Centrifuge Tubes for QuEChERS | |
| QuEChERS Sample Prep | |
| 500 |