missing translation for 'onlineSavingsMsg'
Learn More

CDS Empore™ SDB-RPS Extraction Disks

Catalog No. p-7162246
Change view
Click to view available options
Diameter (Metric) Outer:
4.7 cm
9.0 cm
Quantity:
30/Cs.
60/Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Outer Quantity
13110022 4.7 cm 60/Cs.
13110023 9.0 cm 30/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13110022 Supplier CDS Supplier No. 98060402264EA
Only null left

Formerly 3M™ Empore™ SDB-RPS Solid Phase Extraction Disks (SPE): 2241 & 2341. Reduce extraction time and solvent use for analytical preparation of aqueous environmental samples.

  • Approved for use in EPA Method 515.2
  • Used to collect the more water-soluble analytes from water samples
  • Adsorbent is effective for explosives, pesticides, phenolics, and drugs of abuse
  • Provide up to 90% solvent reduction, excellent processing speed, and contamination-free extractions with accurate and reproducible results
  • Available with a variety of packings in 47 and 90mm sizes

Specifications

Certifications/Compliance EPA Method 515.2
Column Phase Reverse
For Use With (Application) Analytical Preparation of Aqueous Environmental Samples
Sorbent SDB-RPS
Type Extraction Disk
Quantity 60/Cs.
Solvent Reduction 90%
Length (Metric) 4.7 cm
Diameter (Metric) Outer 4.7 cm