missing translation for 'onlineSavingsMsg'
Learn More

CDS Empore™ Carbon Extraction Disks

Catalog No. p-7162248
Change view
Click to view available options
Diameter (Metric) Outer:
4.7 cm
9.0 cm
Quantity:
30/Cs.
60/Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Outer Quantity
13110027 4.7 cm 60/Cs.
13110028 9.0 cm 30/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13110027 Supplier CDS Supplier No. 98060402355EA
Only null left

Formerly 3M™ Empore™ Carbon Solid Phase Extraction Disks (SPE): 2272 & 2372. Provide all the advantages of activated carbon in a compact and thin disk.

  • Designed for the extraction of volatile, highly polar, and water soluble compounds from aqueous solutions
  • Available with a variety of packings in 47 and 90 mm sizes

Specifications

Base Material Activated Carbon
For Use With (Application) Extraction of Volatile, Highly Polar, and Water Soluble Compounds From aq. Soln.
Type Activated Carbon Extraction Disk
Quantity 60/Cs.
Length (Metric) 4.7 cm
Diameter (Metric) Outer 4.7 cm