Learn More
Emodin
A protein tyrosine kinase inhibitor | CAS: 518-82-1 | C15H10O5 | 270.24 g/mol
Supplier: Thermo Scientific Chemicals J61600MC
Description
A protein tyrosine kinase inhibitor
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Emodin | |
| Orange | |
| MFCD00001207 | |
| 1888141 | |
| emodin, schuttgelb, emodol, frangula emodin, rheum emodin, frangulic acid, 3-methyl-1,6,8-trihydroxyanthraquinone, archin, persian berry lake, 1,3,8-trihydroxy-6-methyl-9,10-anthraquinone | |
| CC1=CC(=C2C(=C1)C(=O)C3=CC(=CC(=C3C2=O)O)O)O | |
| 270.24 | |
| CHEBI:42223 | |
| Powder |
| 518-82-1 | |
| C15H10O5 | |
| 100 mg | |
| 14,3561 | |
| RHMXXJGYXNZAPX-UHFFFAOYSA-N | |
| 1,3,8-trihydroxy-6-methylanthracene-9,10-dione | |
| 3220 | |
| 270.24 |
Chemical Identifiers
| 518-82-1 | |
| 270.24 | |
| RHMXXJGYXNZAPX-UHFFFAOYSA-N | |
| 3220 | |
| 1,3,8-trihydroxy-6-methylanthracene-9,10-dione |
| C15H10O5 | |
| MFCD00001207 | |
| emodin, schuttgelb, emodol, frangula emodin, rheum emodin, frangulic acid, 3-methyl-1,6,8-trihydroxyanthraquinone, archin, persian berry lake, 1,3,8-trihydroxy-6-methyl-9,10-anthraquinone | |
| CHEBI:42223 | |
| CC1=CC(=C2C(=C1)C(=O)C3=CC(=CC(=C3C2=O)O)O)O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 208-258-8
RTECSNumber : CB7920600
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only