missing translation for 'onlineSavingsMsg'
Learn More

Emodin 96.0+%, TCI America™

Catalog No. p-7129033
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0500100MG 100 mg
E05001G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0500100MG Supplier TCI America Supplier No. E0500100MG
Only null left

Chemical Identifiers

CAS 518-82-1
Molecular Formula C15H10O5
Molecular Weight (g/mol) 270.24
MDL Number MFCD00001207
InChI Key RHMXXJGYXNZAPX-UHFFFAOYSA-N
Synonym emodin, schuttgelb, emodol, frangula emodin, rheum emodin, frangulic acid, 3-methyl-1,6,8-trihydroxyanthraquinone, archin, persian berry lake, 1,3,8-trihydroxy-6-methyl-9,10-anthraquinone
PubChem CID 3220
ChEBI CHEBI:42223
IUPAC Name 1,3,8-trihydroxy-6-methylanthracene-9,10-dione
SMILES CC1=CC(=C2C(=C1)C(=O)C3=CC(=CC(=C3C2=O)O)O)O

Specifications

Melting Point 253°C
Quantity 100 mg
Formula Weight 270.24
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Emodin
RTECSNumber CB7920600
TSCA No