missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Emodin, 95%, tech.

Chemical Identifiers

CAS 518-82-1
Molecular Formula C15H10O5
Molecular Weight (g/mol) 270.23
MDL Number MFCD00001207
InChI Key RHMXXJGYXNZAPX-UHFFFAOYSA-N
Synonym emodin, schuttgelb, emodol, frangula emodin, rheum emodin, frangulic acid, 3-methyl-1,6,8-trihydroxyanthraquinone, archin, persian berry lake, 1,3,8-trihydroxy-6-methyl-9,10-anthraquinone
PubChem CID 3220
ChEBI CHEBI:42223
IUPAC Name 1,3,8-trihydroxy-6-methylanthracene-9,10-dione
SMILES CC1=CC(=C2C(=C1)C(=O)C3=CC(=CC(=C3C2=O)O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC117751000
SDS View Documents
Thermo Scientific™
117751000
100mg Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC117755000
SDS View Documents
Thermo Scientific™
117755000
500mg Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

518-82-1
270.23
RHMXXJGYXNZAPX-UHFFFAOYSA-N
3220
1,3,8-trihydroxy-6-methylanthracene-9,10-dione
C15H10O5
MFCD00001207
emodin, schuttgelb, emodol, frangula emodin, rheum emodin, frangulic acid, 3-methyl-1,6,8-trihydroxyanthraquinone, archin, persian berry lake, 1,3,8-trihydroxy-6-methyl-9,10-anthraquinone
CHEBI:42223
CC1=CC(=C2C(=C1)C(=O)C3=CC(=CC(=C3C2=O)O)O)O

Specifications

518-82-1
94% min. (HPLC)
C15H10O5
100mg
15, 3618
RHMXXJGYXNZAPX-UHFFFAOYSA-N
1,3,8-trihydroxy-6-methylanthracene-9,10-dione
3220
270.23
Technical
Authentic
Glass bottle
MFCD00001207
08, 520
emodin, schuttgelb, emodol, frangula emodin, rheum emodin, frangulic acid, 3-methyl-1,6,8-trihydroxyanthraquinone, archin, persian berry lake, 1,3,8-trihydroxy-6-methyl-9,10-anthraquinone
CC1=CC(=C2C(=C1)C(=O)C3=CC(=CC(=C3C2=O)O)O)O
270.23
CHEBI:42223
95%
Emodin, tech., 95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 208-258-8

RTECSNumber : CB7920600

SDS
Documents

Documents

RUO – Research Use Only