missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poltex Emergency Airway Procedure Storage Box
Storage emergency crash box designed to hold resuscitation bag and tubing, manual ventilators, etc. Magnetic closure, stainless steel hinges and a front latch accepts either a locking safety seal or padlock.
$380.48 - $485.45
Specifications
| Product Type | Emergency Airway Procedure Storage Box |
|---|---|
| Color | Clear |
| Material | PETG |
| For Use With (Equipment) | Resuscitation Bag and Tubing |
| Catalog Number | Mfr. No. | Dimensions (L x W x H) | Style | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Dimensions (L x W x H) | Style | Price | Quantity & Availability | ||||
|
22022596
|
Poltex
AMBUBAG14714M |
7 x 14 x 14 in. | Magnetic Mount |
Each for $432.96
|
|
||||
|
22022595
|
Poltex
AMBUBAG14714T |
7 x 14 x 14 in. | Tape Mount |
Each for $427.72
|
|
||||
|
22022594
|
Poltex
AMBUBAG14714W |
7 x 14 x 14 in. | Wall Mount |
Each for $380.48
|
|
||||
|
22022593
|
Poltex
AMBUBAG16820M |
8 x 16 x 20 in. | Magnetic Mount |
Each for $485.45
|
|
||||
|
22022592
|
Poltex
AMBUBAG16820T |
8 x 16 x 20 in. | Tape Mount |
Each for $480.19
|
|
||||
|
22022591
|
Poltex
AMBUBAG16820W |
8 x 16 x 20 in. | Wall Mount |
Each for $432.96
|
|
||||
Description
- Emergency crash box holds resuscitation bag and tubing, manual ventilators, etc.
- Magnetic closure and stainless steel hinges
- Front latch accepts either a locking safety seal, or a padlock
- Made from clear PETG
- Choose from two sizes
- Available as magnet mount (powerful magnets included), wall mount (screws/anchors included), and tape mount (VHB tape included)
Specifications
| Emergency Airway Procedure Storage Box | |
| Clear | |
| PETG | |
| Resuscitation Bag and Tubing |