Learn More
Pyramex™ Emerge™ Plus Safety Eyewear
Catalog No. 19155942
Change view
Click to view available options
Product Type:
Full Reader
Top Insert Reader
Magnification Power:
+2.00
+3.00
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Magnification Power
19155942 Full Reader +3.00
19155944 Top Insert Reader +2.00
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19155942 Supplier Pyramex™ Supplier No. SG9810R30
Emerge™ Plus Safety Eyewear features a clear polycarbonate lens with nylon frame.
  • Rubber temple tips
  • Soft nosepiece provides comfort for extended use
  • Translucent grey frame
  • Available in full readers or top insert readers
Specifications
Certifications/Compliance ANSI Z87.1, CSA Z94.3-07
Lens Coating Scratch-resistant, UV-treated
Lens Tint Clear
Product Type Full Reader
Frame Color Translucent Gray
Magnification Power +3.00
Weight (Metric) 31.3 g
Disposable Reusable
Lens Material Polycarbonate
Frame Material Nylon
UV Protection U6
Frame Type Full Frame
Show More Show Less