Learn More
Pyramex™ Emerge™ Plus Safety Eyewear
Catalog No. 19155945
Change view
Click to view available options
Product Type:
Top Insert Reader
Magnification Power:
+2.50
Catalog No. Product Type Magnification Power
19155945 Top Insert Reader +2.50
1 options
Catalog No. 19155945 Supplier Pyramex™ Supplier No. SG9810TR25
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Emerge™ Plus Safety Eyewear features a clear polycarbonate lens with nylon frame.
  • Rubber temple tips
  • Soft nosepiece provides comfort for extended use
  • Translucent grey frame
  • Available in full readers or top insert readers
Specifications
Lens Coating Scratch-resistant, UV-treated
Certifications/Compliance ANSI Z87.1, CSA Z94.3-07
Description The top insert reader is strategically placed for applications in which user is frequently looking up
Lens Tint Clear
Product Type Top Insert Reader
Frame Color Translucent Gray
Magnification Power +2.50
Weight (Metric) 31.3 g
Disposable Reusable
Lens Material Polycarbonate
Frame Material Nylon
UV Protection U6
Frame Type Full Frame
Show More Show Less