Learn More
Superior Glove™ Emerald CX™ Nylon/Stainless Steel Knit Cut-Resistant Gloves
Nylon and fine-gauge steel cut-resistant gloves with polyurethane palm coating provides a secure grip, dexterity and touch sensitivity.
$8.74 - $8.74
Specifications
ANSI Abrasion Rating ANSI 4
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A2
Material Nylon, Stainless Steel Fiber
View More Specs
Products 7
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
29-636-1681
View Documents
Superior Glove™
S13KBPU6
6
Pair for $8.74
Only null left
29-636-1682
View Documents
Superior Glove™
S13KBPU7
7
Pair for $8.74
Only null left
29-636-1683
View Documents
Superior Glove™
S13KBPU8
8
Pair for $8.74
Only null left
29-636-1684
View Documents
Superior Glove™
S13KBPU9
9
Pair for $8.74
Only null left
29-636-1678
View Documents
Superior Glove™
S13KBPU10
10
Pair for $8.74
Only null left
29-636-1679
View Documents
Superior Glove™
S13KBPU11
11
Pair for $8.74
Only null left
29-636-1680
View Documents
Superior Glove™
S13KBPU12
12
Pair for $8.74
Only null left
Description
Description
  • Combination of nylon and fine-gauge steel
  • Lint-free nylon is lightweight and tough for better dexterity
  • Comfortable with touch sensitivity
  • Polyurethane palm coating provides secure grip and tactile feel
  • Ideal for glass handling, non-marring gloves will not leave residue, smudges or fingerprints
  • Extra-long knit wrists provide extra protection
  • Touchscreen compatible
Specifications
Specifications
ANSI 4
Gray
Nylon, Stainless Steel Fiber
HVAC, automotive, sharp Steel, Glass, Metal fabrication, light Metal stamping
ANSI/ISEA 105-16 Level 2
13
Polyurethane
ANSI A2
Cut-resistant Gloves
Knit
Emerald CX™
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'