Learn More
MilliporeSigma™ Water Detector for Water Purification Systems
Catalog No. ZFWATDET1
Change view
Click to view available options
120V
Catalog No.
ZFWATDET1 120V
1 options
Catalog No. ZFWATDET1 Supplier MilliporeSigma™ Supplier No. ZFWATDET1
Detects water spillage on the floor and automatically closes the stand-alone inlet solenoid valve to protect the surroundings from water damage
Water purification accessory
Specifications
Type Detector
For Use With (Equipment) Millipore™ Pure Water Storage Reservoir
Voltage 120 V