missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Vesicular Glutamate Transporter 2 Control Peptide

Catalog No. AG209MI
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AG209MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AG209MI Supplier MilliporeSigma™ Supplier No. AG209

Specifications

Gene ID (Entrez) NM_020346.1
Concentration 1 mg/mL
Content And Storage -20°C in undiluted aliquots for up to 3 months. Avoid repeated freeze/thaw cycles.
Format Liquid
Formulation Liquid
Gene Symbol SLC17A6, DNPI, VGLUT2
Product Type Vesicular Glutamate Transporter 2, control peptide for AB5907
Quantity 100 μg
Regulatory Status RUO