missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Vesicular Glutamate Transporter 1 Control Peptide
Specifications
Specifications
| Gene ID (Entrez) | NP_446311 |
| Concentration | 1 mg/mL |
| Content And Storage | -20°C in undiluted aliquots for up to 3 months. Avoid repeated freeze/thaw cycles. |
| Format | Liquid |
| Formulation | Liquid |
| Gene Symbol | VGluT1, Solute carrier family 17 member 7, Brain-specific Na(+)-dependent inorganic phosphate cotransporter, Slc17a7, Bnpi |
| Product Type | Vesicular Glutamate Transporter 1, control peptide for AB5905 |
| Quantity | 100 μg |
| Regulatory Status | RUO - research use only |