Learn More
MilliporeSigma™ UV Lamps and Assemblies for Water Purification Systems
Control bacteria and oxidize organics
Supplier: MilliporeSigma™ SIM0000UV
Description
Control bacteria and oxidize organics
- Replacement parts for EMD Millipore Water Purification Systems and A10 TOC Monitor
Specifications
| UV Lamp | |
| Synergy UV, Simplicity UV | |
| 185 nm |