Learn More
MilliporeSigma™ UV Lamps and Assemblies for Water Purification Systems
Control bacteria and oxidize organics
Supplier: MilliporeSigma™ ZFA10UV01
Description
Control bacteria and oxidize organics
- Replacement parts for EMD Millipore Water Purification Systems and A10 TOC Monitor
Chemical Identifiers
| UV Lamp |
Specifications
| UV Lamp | |
| A10 TOC Monitor (external), Milli-Q Ultrapure A10, High-volume Elix TOC Upgrade Water Purification Kit | |
| 254/185 nm |