Learn More
MilliporeSigma™ UV Lamp for Laboratory Water Systems
For destruction of organic contaminants in water by photo-oxidation.
Supplier: MilliporeSigma™ SYN185UV1
Description
For destruction of organic contaminants in water by photo-oxidation
- For use with Direct-Q 3 UV, Simplicity UV, and RiOs DI 3 UV systems built since 2003
- Wavelength: 185nm
Greener Choice Claims
- Environmental benefits include:
- Recyclable
Specifications
UV Lamp | |
Direct-Q™ 3 UV, Simplicity UV System | |
185 nm |