missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Upstate™ Ser/Thr Phosphatase Assay Kit 1 (K-R-pT-I-R-R)

Catalog No. 17127
Change view
Click to view available options
Quantity:
1 Kit (100 Assays)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17127 1 Kit (100 Assays)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17127 Supplier MilliporeSigma™ Supplier No. 17127

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Phosphatase Assay

Specifications

Description Ser/Thr Phosphatase Assay Kit 1 (K-R-pT-I-R-R)
Detection Method Colorimetric
For Use With (Application) Phosphatase Assay
Includes 96-well microtiter plate, Malachite Green Solution A, Malachite Green Additive, Phosphate Standard, Threonine Phosphopeptide (K-R-pT-I-R-R), pNPP (p-Nitrophenyl Phosphate), pNPP Ser/Thr Assay Buffer, NiCl2, 40mM
Quantity 1 Kit (100 Assays)