missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Upstate Rac1/Cdc42 Activation Assay Kit

Catalog No. 17441
Change view
Click to view available options
Quantity:
30 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17441 30 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17441 Supplier MilliporeSigma™ Supplier No. 17441

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Activity Assay, Affinity Binding Assay, G-Protein Assay

  • Uses a PAK1 binding domain coupled to agarose beads.
  • Isolates active GTP-bound Rac1 and Cdc42 for immunoblot detection.

Specifications

Content And Storage -20°C for 12 months
Description ac1/Cdc42 Activation Assay Kit
Detection Method Chemiluminescence
For Use With (Application) Activity Assay, Affinity Binding Assay, G-Protein Assay
Includes 100X GDP (100mM), Rac/cdc42 Assay Reagent, Anti-Rac1 (clone 23A8), Anti-cdc42, (mouse monoclonal IgG1), Mg2+ Lysis/Wash Buffer (5X), 100X GTPyS (10mM)
Quantity 30 Assays
Type Activation Assay Kit