missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Upstate™ PTP Assay Kit 2

Catalog No. 17126
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17126 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17126 Supplier MilliporeSigma™ Supplier No. 17126

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Phosphatase Assay

Specifications

Description PTP Assay Kit 2
Detection Method Colorimetric
For Use With (Application) Phosphatase Assay
Includes 96-well microtiter plate, Tyrosine Phosphopeptide (TSTEPQpYQPGENL), Malachite Green Solution A, Malachite Green Additive, Phosphate Standard, pNPP (p-Nitrophenyl Phosphate), pNPP Tyr Assay Buffer
Quantity 1 Kit
Type Activity Assay Kit