missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Upstate™ PTP Assay Kit 1
Description
- Supports phosphopeptide dephosphorylation with malachite green detection.
- Also supports pNPP hydrolysis.
Specifications
Specifications
| Description | PTP Assay Kit 1 |
| Detection Method | Colorimetric |
| For Use With (Application) | Phosphatase Assay |
| Includes | Malachite Green Additive, Phosphate Standard, pNPP (p-Nitrophenyl Phosphate), pNPP Tyr Assay Buffer, 96-well microtiter plate, Tyrosine Phosphopeptide, Malachite Green Solution A |
| Quantity | 1 Kit (100 Assays) |
| Type | Activity Assay Kit |