missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Upstate Mouse NGF 2.5S Protein

Catalog No. 01125MI
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01125MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01125MI Supplier MilliporeSigma™ Supplier No. 01125

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The mouse NGF 2.5S Protein( or nerve growth factor 2.5S) is available in a 100μg format.

The protein encoded by this gene is a member of the neurotrophin family, that controls survival and differentiation of mammalian neurons. This protein is closely related to both nerve growth factor and brain-derived neurotrophic factor. It may be involved in the maintenance of the adult nervous system, and may affect development of neurons in the embryo when it is expressed in human placenta. NTF3-deficient mice generated by gene targeting display severe movement defects of the limbs. The mature peptide of this protein is identical in all mammals examined including human, pig, rat and mouse.

Specifications

For Use With (Application) Cell Culture
Quantity 100 μg
Source Male mouse submaxillary glands