missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Upstate™ CpGenome™ Human Non-Methylated DNA Standard Set

Catalog No. S8001U
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S8001U 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S8001U Supplier MilliporeSigma™ Supplier No. S8001U

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For gene methylation studies

Specifications

Content And Storage -20°C for 6 months
Description CpGenome human non-methylated DNA standard set
For Use With (Application) Gene methylation studies
Quantity 5 μg