missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Universal pH Indicator Papers, pH 1 to 14

Catalog No. M1109620003
Change view
Click to view available options
Quantity:
3 Rolls
6 Rolls
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1109620003 3 Rolls
M1102320001 6 Rolls
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1109620003 Supplier MilliporeSigma™ Supplier No. 1.10962.0003

Since they protect against external influences, pH indicator papers supplied in rolls are well suited for long-term storage

  • High-quality filter paper impregnated with indicator solutions
  • M1102320001 are replacement rolls for M1109620003
  • Color scale: graduated by 1 pH units

Specifications

pH Range 1 to 14
For Use With (Application) Environmental analysis and industrial in-process controls, calibration and checking of the products by certified buffer solutions, rapid on-the-spot pH determination
Graduations 1-2-3-4-5-6-7-8-9-10-11-12-13-14
Quantity 3 Rolls
Storage Requirements 10°C to 25°C
Type Universal pH Indicator Paper