Learn More
MilliporeSigma™ Universal pH Indicator Papers, pH 1 to 14
Since they protect against external influences, pH indicator papers supplied in rolls are well suited for long-term storage
Supplier:  MilliporeSigma™ 1.10232.0001
 View more versions of this product
Catalog No. M1102320001
Only null left
Description
Description
  • High-quality filter paper impregnated with indicator solutions
  • M1102320001 are replacement rolls for M1109620003
  • Color scale: graduated by 1 pH units
Note: these are replacement rolls only; no chart is included.
Specifications
Chemical Identifiers
1 to 14
Environmental analysis and industrial in-process controls, calibration and checking of the products by certified buffer solutions, rapid on-the-spot pH determination
10°C to 25°C
Universal pH Indicator Paper
Specifications
1 to 14
6 replacement rolls. (No chart included.)
10°C to 25°C
Environmental analysis and industrial in-process controls, calibration and checking of the products by certified buffer solutions, rapid on-the-spot pH determination
6 Rolls
Universal pH Indicator Paper
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
ShelfLife : 3 to 5 Years