Learn More
MilliporeSigma™ TLC Silica Gel 60 Aluminum Sheets
For thin layer chromatography applications.
Supplier:  MilliporeSigma™ 1055530001
 View more versions of this product
Catalog No. M1055530001
Only null left
Explore available promotions
Description
Description
Our classical silica TLC plates are based on a combination of our proven silica gel 60 and the addition of a unique polymeric binder resulting in a very adherent and hard surface that will not crack or blister and even allow writing with a pensil on the surface without risk to damage the layer.
Our concentrating zone plates are based on different adsorption properties of two adsorbents. The first is a large pore concentrating adsorbent where the samples are applied; the second is a selective layer for separation. Regardless of the spots’ shape, size, or position, the sample always concentrates as a narrow band where the two adsorbents overlap and where the separation starts. These plates allow users to quickly and easily apply any kind of sample, even large volumes of diluted samples.
Specifications
Specifications
20 to 200 min.
10 to 12 μm
25/Pk.
In-Process Control in Drug Synthesis, Identity and Stability Testing of Drugs, Quality Control of Pharmaceuticals, Food and Environmental Samples, Residue Analysis in Food and Environmental Samples
5 to 20 μm
Silica Gel 60
200 μm
20 cm
0.0011 in.
Aluminum
60 Å
Classical Silica TLC Plate
10.0
0.8 mL/g
520 m2/g
20 x 20 cm
20 cm
30 μm
Product Suggestions
Product Suggestions