Learn More
MilliporeSigma™ Synuclein α Control Peptide
Supplier: MilliporeSigma™ AG240
Specifications
| NM_000345.2, NM_007308.1 | |
| Lyophilized | |
| SNCA, alpha-synuclein, NACP, MGC110988, PARK1, PARK4, Alpha-synuclein, PD1 | |
| 100 μg |
| Maintain lyophilized material at 2-8°C for up to 6 months. After reconstitution -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles. | |
| Lyophilized. Reconstitute with 100μL of sterile distilled water. Centrifuge to remove insoluble material. Contains no preservative. | |
| Synuclein α, control peptide for AB5334P | |
| RUO - research use only |