missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Synergypak 1 Cartridge

Catalog No. SYPK0SIA1
Change view
Click to view available options
Pore Size:
0.22 μm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size
SYPK0SIA1 0.22 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SYPK0SIA1 Supplier MilliporeSigma™ Supplier No. SYPK0SIA1
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

With ion-exchange technology

SynergyPak, one Purification Cartridge for Synergy™ systems connected to Elix™ / RO / distilled water feed

Specifications

Feedwater Source Elix, Reverse Osmosis, Distilled
Material Activated Carbon
Pore Size 0.22 μm
Purification Method Ion Exchange
For Use With (Equipment) Synergy systems connected to Elix/RO/distilled water feed
Model SynergyPak 1
Type Synergy™ Pak 1 Purification Cartridge