missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Swab in Vial

Catalog No. MMSB10025
Change view
Click to view available options
Applicator Material:
Polyethylene
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Applicator Material
MMSB10025 Polyethylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MMSB10025 Supplier MilliporeSigma™ Supplier No. MMSB10025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Self-contained unit with Dacron™ swab suspended in phosphate buffer

  • Polyethylene paddle with polyethylene cap
  • Clear vial for easy viewing of contents
  • SAN chamber

Specifications

Applicator Material Polyethylene
For Use With (Application) Clinical lab, environmental monitoring, food and beverage, water monitoring applications
Color Clear
Product Type Swab