missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Super-Q™ Plus Water Purification System Accessories

For Super-Q Systems

$875.05 - $1,490.00

Specifications

For Use With (Equipment) Water Purification System
Capacity (English) 22 in.
Capacity (Metric) 56 cm
Length (English) 22 in.
Length (Metric) 56 cm
View More Specs

Products 2
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
CDFC02203
View Documents Promotion Details
MilliporeSigma™
CDFC02203
Cartridge, Ion Exchange
Pack of 3 Each for $1,490.00
Only null left
 
CPEX02201
View Documents Promotion Details
MilliporeSigma™
CPEX02201
Organex-Q Cartridge
Each for $875.05
Only null left
 
Description

Description

For Super-Q systems
Specifications

Specifications

Water Purification System
56 cm
56 cm
22 in.
22 in.
Videos
SDS
Documents

Documents