missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Spill Kit, multipurpose, SpillSolv

Catalog No. MSX13001
Change view
Click to view available options
Spill Kit, Multipurpose
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MSX13001 Spill Kit, Multipurpose
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MSX13001 Supplier MilliporeSigma™ Supplier No. SX13001
Only null left

Complete kit includes reagents, splash goggles with ventilation, nitril gloves, disposal bags, plastic scoops and scrapers, sponge for residue clean up and treatment guide

Specifications

Type Multipurpose Spill Kit
Includes 2 x 1 kg SpillSolv acid neutralizer, 2 x 0.9 kg SpillSolv caustic neutralizer, 2 x 0.5 kg SpillSolv volatile adsorbent powder, 1 pair splash goggles with ventilation, 1 pair nitrile gloves A15
Dimensions (L x W x H) 1.58 x 7 x 13 in. (4.01 x 177.8 x 33.02 cm)
Length (English) 1.58 in.
Length (Metric) 4.01 cm
Width (English) 7 in.
Width (Metric) 177.8 cm
Height (English) 13 in.
Height (Metric) 33.02 cm