Learn More
MilliporeSigma™ SNAP i.d.™ 2.0 MultiBlot Holding Frames and Holders
Catalog No. SNP2BHMB050
Change view
Click to view available options
Type:
MultiBlot Holder
Quantity:
50/Pk.
Catalog No. Type Quantity
SNP2BHMB050 MultiBlot Holder 50/Pk.
1 options
Catalog No. SNP2BHMB050 Supplier MilliporeSigma™ Supplier No. SNAP2BHMB050
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For SNAP i.d.™ 2.0 System
Multiblot Holder
  • Each half blot membrane is aligned horizontally, allowing for uniform distribution of antibody
  • Flat profile allows for compact space allocation for storage, shipping and waste management
  • Size: 4.5 x 8.4cm
Multiblot Holding frame
  • Stackable frame with lid, easy locking latch and umbrella valve for offline incubation
  • Clear lids retains humidity
Western Blotting
Specifications
Type MultiBlot Holder
Quantity 50/Pk.