Learn More
MilliporeSigma™ SNAP i.d.™ 2.0 MultiBlot Holding Frames and Holders
For SNAP i.d.™ 2.0 System
Supplier:  MilliporeSigma™ SNAP2FRMB01
 Afficher plus de versions de ce produit
Numéro de catalogue. SNAP2FRMB01
Il en reste null
Description
Description
Multiblot Holder
  • Each half blot membrane is aligned horizontally, allowing for uniform distribution of antibody
  • Flat profile allows for compact space allocation for storage, shipping and waste management
  • Size: 4.5 x 8.4cm
Multiblot Holding frame
  • Stackable frame with lid, easy locking latch and umbrella valve for offline incubation
  • Clear lids retains humidity
Western Blotting
Spécifications
Spécifications
MultiBlot Holding Frame
Single Pack
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS