missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ SMARTPak Purification Pack for RiOs 5 and 8 Systems

For use with RiOs™ 8 Systems

Supplier:  MilliporeSigma™ SPR08S001

Catalog No. SPR08S001


Only null left

Description

Description

For use with RiOs™ 8 Systems

SmartPak™ RO 8 Purification Pack
Specifications

Specifications

RiOs™ 5 and 8 system
Purification Pack
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions