missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ SimpliPak™ 2 Cartridge for Simplicity™ Ultrapure Water Purification System

Catalog No. SIPK0SIX2
Change view
Click to view available options
Purification Method:
Ion Exchange
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Purification Method
SIPK0SIX2 Ion Exchange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SIPK0SIX2 Supplier MilliporeSigma™ Supplier No. SIPK0SIX2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Produces ultrapure water from a variety of pre-treated water types

  • Based on “Ion-exchange” technology
  • Snap-in-and-out design for easy and rapid replacement

Specifications

Feedwater Source Milli-DI, Deionized
Purification Method Ion Exchange
For Use With (Equipment) Simplicity™ Ultrapure Water Purification System
Model SimpliPak 2