Learn More
MilliporeSigma™ Silica Gel 60 F254 PLC Plates
Glass plates 20 x 20 cm
Supplier: MilliporeSigma™ 1.05717.0001
Description
PLC plates allow users to separate samples that vary greatly in size – from grams down to milligrams. Available with or without a fluorescence indicator, PLC plates are available in 0.5mm, 1mm, and 2mm thickness. They also use our same proven silica-binder technology as in analytical TLC plates. To isolate the substance by extraction, users can simply scrape the spot from the layer.Chemical Identifiers
| 20 x 20 cm |
Specifications
| 12/Pk. | |
| Layer: 2 mm | |
| 20 x 20 cm |