Learn More
MilliporeSigma™ Silica Gel 60 F254 + 366 Coated Glass-Backed PLC Plates
allow users to separate samples that vary greatly in size – from grams down to milligrams
Supplier: MilliporeSigma™ 1.05637.0001
Description
Available with or without a fluorescence indicator, PLC plates has 2mm thickness. They also use our same proven silica-binder technology as in analytical TLC plates. To isolate the substance by extraction, users can simply scrape the spot from the layer.Specifications
| Glass | |
| Silica PLC Plate | |
| Highly Suitable for a Variety of Preparative AppliCatIons Including Cleaning and Enrichment of Synthetic Reaction Mixtures, Natural Products, Plant Extracts and Biotechnological Products | |
| 2 mm | |
| 20 cm |
| 12/Pk. | |
| 254 and 366 nm | |
| Silica Gel 60 F254 + 366 | |
| 20 x 20 cm | |
| 20 cm |