missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ S-100b Protein, Bovine Brain, Protein

Catalog No. 5592901MG
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5592901MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5592901MG Supplier MilliporeSigma™ Supplier No. 5592901MG
Only null left
Native homodimeric S-100b isoform composed of ββ-subunits, purified from bovine brain. Elevated serum level of S-100b is used as a marker of stroke and cerebral edema.

Specifications

Formulation Lyophilized from 50mM Tris-HCl, pH 7.5, 50mM NaCl, 100μM DTT.
Preparation Method 50mM Tris buffer, pH 8.0 (5mg/mL)
Purification Method SDS-PAGE
Quantity 1 mg
Immunogen S-100b Protein
Storage Requirements Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 6 months at −20°C.
Regulatory Status RUO
Species Bovine
Recombinant Native
Form Lyophilized
Purity or Quality Grade ≥95%
Show More Show Less