missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ ROCK Inhibitor (Y-27632)

Catalog No. scm075
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCM075 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCM075 Supplier MilliporeSigma™ Supplier No. SCM075
Only null left

available in a 5 mg format and has been optimized and validated for cell culture and Neuroscience applications.

Specifications

Content And Storage -20°C
For Use With (Application) Neuroscience and cell culture
Quantity 5 mg