missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Reflectoquant™ RQflex™ plus Potassium Test Strips

Catalog No. M1179450001
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1179450001 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1179450001 Supplier MilliporeSigma™ Supplier No. 1.17945.0001
Only null left

Includes

Reagent, if needed

Used in reflectometric test

  • With Kalignost™, potassium can be measured as turbidity reflectometrically in drinking water, groundwater and surface water
  • Special applications for determination in soil are available

Range: 1.0 to 25.0mg/L

Order Info

Pack of 100 tests

Specifications

For Use With (Application) Can be measured as turbidity reflectometrically in drinking water, groundwater and surface water
For Use With (Equipment) Reflectometric
Graduations 1 to 25mg/L
Quantity 100 Tests
Range 1 to 25 mg/L
Storage Requirements 15°C to 25°C
Test Method Kalignost, turbidimetric
Type Potassium Test Strip